siRNA Details
Gene Symbol: Fgf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130753.1 | NP_570109.1 | 1, 2 | 245 | ||
GenBank mRNA | AB078900.1 | - | 1, 2 | 245 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fgf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130753.1 | NP_570109.1 | 1, 2 | 245 | ||
GenBank mRNA | AB078900.1 | - | 1, 2 | 245 |