siRNA Details
| Gene Symbol: Atp2c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131907.2 | NP_571982.2 | 6, 7 | 835 | ||
| GenBank mRNA | BC086994.1 | - | 6, 7 | 835 | 7106233 | |
| M93017.1 | - | 6, 7 | 546 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001253836, NM_001253834, NM_175025, NM_001253831, AK133703, and others.

Atp2c1
Genomic Map