siRNA Details
| Gene Symbol: As3mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080890.1 | NP_543166.1 | 9 | 899 | ||
| GenBank mRNA | AF393243.1 | - | 9 | 899 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

As3mt| Gene Symbol: As3mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080890.1 | NP_543166.1 | 9 | 899 | ||
| GenBank mRNA | AF393243.1 | - | 9 | 899 | ||