siRNA Details
| Gene Symbol: Pde7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080894.2 | NP_543170.1 | 10 | 1006 | ||
| XM_008758591.1 | XP_008756813.1 | 9 | 1326 | |||
| XM_008758592.1 | XP_008756814.1 | 9 | 910 | |||
| GenBank mRNA | AB057409.1 | - | 10 | 1006 | ||
| AB057410.1 | - | 9 | 874 | |||
| AB057411.1 | - | 11 | 975 | |||

Pde7b
Genomic Map