siRNA Details
| Gene Symbol: Ppp4r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080907.2 | NP_543183.1 | 5, 6 | 789 | ||
| GenBank mRNA | AF332004.1 | - | 5, 6 | 707 | ||
| BC061726.1 | - | 5, 6 | 789 | 5598703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp4r1| Gene Symbol: Ppp4r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080907.2 | NP_543183.1 | 5, 6 | 789 | ||
| GenBank mRNA | AF332004.1 | - | 5, 6 | 707 | ||
| BC061726.1 | - | 5, 6 | 789 | 5598703 | ||