siRNA Details
| Gene Symbol: Cfh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130409.2 | NP_569093.2 | 5 | 555 | ||
| XM_008769531.1 | XP_008767753.1 | 5 | 752 | |||
| XM_008769532.1 | XP_008767754.1 | 5 | 751 | |||
| XM_008769533.1 | XP_008767755.1 | 5 | 752 | |||
| GenBank mRNA | AJ320522.1 | - | 529 | |||
| BC083174.1 | - | 564 | 7096787 | |||

Cfh
Genomic Map