siRNA Details
Gene Symbol: Abcg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130414.2 | NP_569098.2 | 7 | 1181 | ||
XM_008764453.1 | XP_008762675.1 | 5 | 1222 | |||
GenBank mRNA | AF351785.2 | - | 7 | 1181 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abcg8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130414.2 | NP_569098.2 | 7 | 1181 | ||
XM_008764453.1 | XP_008762675.1 | 5 | 1222 | |||
GenBank mRNA | AF351785.2 | - | 7 | 1181 |