siRNA Details
Gene Symbol: Ggt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130423.2 | NP_569107.2 | 8 | 1067 | ||
XM_008762300.1 | XP_008760522.1 | 8 | 3692 | |||
XM_008762301.1 | XP_008760523.1 | 8 | 1463 | |||
GenBank mRNA | AF244973.1 | - | 8 | 1050 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
