siRNA Details
| Gene Symbol: Tnfrsf1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130426.4 | NP_569110.1 | 6 | 718 | ||
| GenBank mRNA | AF142499.1 | - | 2 | 205 | ||
| AF420214.1 | - | 6 | 589 | |||
| AF498039.2 | - | 6 | 718 | |||
| AY191268.1 | - | 718 | ||||
| AY191269.1 | - | 718 | ||||
| AY344841.1 | - | 6 | 631 | |||

Tnfrsf1b
Genomic Map