siRNA Details
| Gene Symbol: Lcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130741.1 | NP_570097.1 | 5 | 599 | ||
| GenBank mRNA | BC089053.1 | - | 5 | 661 | 7310185 | |
| X13295.1 | - | 5 | 599 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lcn2| Gene Symbol: Lcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130741.1 | NP_570097.1 | 5 | 599 | ||
| GenBank mRNA | BC089053.1 | - | 5 | 661 | 7310185 | |
| X13295.1 | - | 5 | 599 | |||