siRNA Details
| Gene Symbol: VENTX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014468.3 | NP_055283.1 | 3 | 1396 | ||
| GenBank mRNA | AF068006.1 | - | 3 | 1403 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

VENTX| Gene Symbol: VENTX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014468.3 | NP_055283.1 | 3 | 1396 | ||
| GenBank mRNA | AF068006.1 | - | 3 | 1403 | ||