siRNA Details
| Gene Symbol: P2RY10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324218.1 | NP_001311147.1 | 3 | 944 | ||
| NM_001324221.1 | NP_001311150.1 | 3 | 850 | |||
| NM_001324225.1 | NP_001311154.1 | 4 | 929 | |||
| NM_014499.3 | NP_055314.1 | 4 | 993 | |||
| NM_198333.2 | NP_938147.1 | 2 | 801 | |||
| GenBank mRNA | AK292138.1 | - | 4 | 931 | ||
| AK313218.1 | - | 2 | 742 | |||
| BC051875.1 | - | 4 | 904 | 5212240 | ||

P2RY10
Genomic Map