siRNA Details
| Gene Symbol: Pecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133299.2 | NP_579833.1 | 1 | 188 | ||
| XM_008767180.1 | XP_008765402.1 | 1 | 194 | |||
| GenBank mRNA | AF021854.1 | - | 1 | 189 | ||
| AF099742.1 | - | 1 | 108 | |||
| BC060546.1 | - | 1 | 182 | 6889831 | ||

Pecr
Genomic Map