siRNA Details
| Gene Symbol: Ddx39b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133300.3 | NP_579834.2 | 2 | 310 | ||
| GenBank mRNA | BC080243.1 | - | 2 | 259 | 7100155 | |
| DY309526.1 | - | 2 | 310 | |||
| FQ212901.1 | - | 2 | 296 | |||
| M75168.1 | - | 1 | 215 | |||

Ddx39b
Genomic Map