siRNA Details
Gene Symbol: Olr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133306.2 | NP_579840.2 | 1, 2 | 172 | ||
GenBank mRNA | AB005900.1 | - | 1, 2 | 172 | ||
BC097290.1 | - | 1, 2 | 125 | 7458637 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133306.2 | NP_579840.2 | 1, 2 | 172 | ||
GenBank mRNA | AB005900.1 | - | 1, 2 | 172 | ||
BC097290.1 | - | 1, 2 | 125 | 7458637 |