siRNA Details
| Gene Symbol: Slc40a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133315.2 | NP_579849.2 | 5 | 766 | ||
| GenBank mRNA | AF394785.3 | - | 5 | 711 | ||
| CK364414.1 | - | 4 | 398 | |||
| FQ226522.1 | - | 4 | 426 | |||
| U76714.1 | - | 5 | 781 | |||
| Gene Symbol: LOC100911874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750695.3 | XP_003750743.1 | 1189 | |||

Slc40a1
Genomic Map