siRNA Details
| Gene Symbol: Tesk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133396.2 | NP_596887.1 | 2 | 463 | ||
| XM_006238628.2 | XP_006238690.1 | 2 | 694 | |||
| GenBank mRNA | AB049402.1 | - | 2 | 464 | ||
| BC081737.1 | - | 2 | 505 | |||
| BC128727.1 | - | 2 | 469 | 7111312 | ||

Tesk2
Genomic Map