siRNA Details
Gene Symbol: Ep300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765878.1 | XP_008764100.1 | 2 | 1366 | ||
GenBank mRNA | AB066220.1 | - | 2 | 396 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ep300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765878.1 | XP_008764100.1 | 2 | 1366 | ||
GenBank mRNA | AB066220.1 | - | 2 | 396 |