siRNA Details
| Gene Symbol: Chrna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133420.1 | NP_596911.1 | 4, 5 | 582 | ||
| XM_008770735.1 | XP_008768957.1 | 4, 5 | 424 | |||
| GenBank mRNA | AY574253.1 | - | 3, 4 | 278 | ||
| L10077.1 | - | 4, 5 | 582 | |||

Chrna2
Genomic Map