siRNA Details
| Gene Symbol: Ythdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133423.1 | NP_596914.1 | 2 | 86 | ||
| XM_006250805.2 | XP_006250867.1 | 3 | 438 | |||
| XM_006250806.2 | XP_006250868.1 | 2 | 401 | |||
| XM_006250808.2 | XP_006250870.1 | 2 | 401 | |||
| XM_008769987.1 | XP_008768209.1 | 2 | 1453 | |||
| XM_008769988.1 | XP_008768210.1 | 2 | 1453 | |||
| XM_008769989.1 | XP_008768211.1 | 2 | 1453 | |||
| XM_008769990.1 | XP_008768212.1 | 2 | 1454 | |||
| GenBank mRNA | AF144731.1 | - | 2 | 86 | ||
| D78303.1 | - | 2 | 402 | |||

Ythdc1
Genomic Map