siRNA Details
| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004133.1 | NP_001004133.1 | 1 | 570 | ||
| NM_133440.1 | NP_597684.1 | 1 | 676 | |||
| NM_133441.1 | NP_597685.1 | 6 | 1170 | |||
| NM_201350.1 | NP_958738.1 | 15 | 2019 | |||
| XM_008760068.1 | XP_008758290.1 | 14 | 1930 | |||
| GenBank mRNA | AF060116.1 | - | 1 | 570 | ||
| AF141903.1 | - | 5 | 662 | |||
| AF159048.1 | - | 5 | 680 | |||
| AJ131899.1 | - | 1 | 676 | |||
| AJ249562.1 | - | 6 | 1170 | |||
| AY298755.1 | - | 15 | 2019 | |||

Shank2
Genomic Map