siRNA Details
| Gene Symbol: Sval2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133291.1 | NP_579825.1 | 2 | 176 | ||
| GenBank mRNA | AB051831.1 | - | 2 | 176 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sval2| Gene Symbol: Sval2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133291.1 | NP_579825.1 | 2 | 176 | ||
| GenBank mRNA | AB051831.1 | - | 2 | 176 | ||