siRNA Details
| Gene Symbol: Gata3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133293.1 | NP_579827.1 | 2 | 538 | ||
| XM_006254253.2 | XP_006254315.1 | 3 | 1304 | |||
| XM_008771893.1 | XP_008770115.1 | 3 | 1248 | |||
| XM_008771894.1 | XP_008770116.1 | 3 | 2838 | |||
| GenBank mRNA | AY024364.1 | - | 2 | 538 | ||

Gata3
Genomic Map