siRNA Details
| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133313.2 | NP_579847.1 | 5 | 665 | ||
| GenBank mRNA | AJ301634.1 | - | 5 | 774 | ||
| BC062087.1 | - | 5 | 665 | 5621521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atxn10| Gene Symbol: Atxn10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133313.2 | NP_579847.1 | 5 | 665 | ||
| GenBank mRNA | AJ301634.1 | - | 5 | 774 | ||
| BC062087.1 | - | 5 | 665 | 5621521 | ||