siRNA Details
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133316.1 | NP_579850.1 | 4 | 329 | ||
GenBank mRNA | AF478683.1 | - | 4 | 329 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133316.1 | NP_579850.1 | 4 | 329 | ||
GenBank mRNA | AF478683.1 | - | 4 | 329 |