siRNA Details
| Gene Symbol: Crebbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133381.3 | NP_596872.3 | 2 | 587 | ||
| XM_008767524.1 | XP_008765746.1 | 2 | 717 | |||
| GenBank mRNA | AB066219.1 | - | 2 | 564 | ||
| AY462245.1 | - | 2 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crebbp| Gene Symbol: Crebbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133381.3 | NP_596872.3 | 2 | 587 | ||
| XM_008767524.1 | XP_008765746.1 | 2 | 717 | |||
| GenBank mRNA | AB066219.1 | - | 2 | 564 | ||
| AY462245.1 | - | 2 | 587 | |||