siRNA Details
Gene Symbol: Scpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133383.1 | NP_596874.1 | 3 | 262 | ||
GenBank mRNA | AF330051.1 | - | 3 | 262 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133383.1 | NP_596874.1 | 3 | 262 | ||
GenBank mRNA | AF330051.1 | - | 3 | 262 |