siRNA Details
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133392.1 | NP_596883.1 | 3 | 469 | ||
XM_006244908.2 | XP_006244970.1 | 3 | 1238 | |||
GenBank mRNA | AB070349.1 | - | 3 | 469 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stk17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133392.1 | NP_596883.1 | 3 | 469 | ||
XM_006244908.2 | XP_006244970.1 | 3 | 1238 | |||
GenBank mRNA | AB070349.1 | - | 3 | 469 |