siRNA Details
Gene Symbol: Clcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133414.1 | NP_596905.1 | 3 | 215 | ||
GenBank mRNA | AB052921.1 | - | 2 | 173 | ||
AB052922.1 | - | 3 | 215 | |||
AB052923.1 | - | 3 | 235 | |||
AF117330.1 | - | 3 | 203 |
