siRNA Details
| Gene Symbol: Syf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133417.1 | NP_596908.1 | 3 | 267 | ||
| GenBank mRNA | AF366369.1 | - | 3 | 267 | ||
| BC081735.1 | - | 3 | 250 | |||
| BC097289.1 | - | 3 | 275 | 7445525 | ||
| FQ234066.1 | - | 1 | 94 | |||

Syf2
Genomic Map