siRNA Details
| Gene Symbol: Hrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133428.2 | NP_596919.1 | 2, 3 | 340 | ||
| GenBank mRNA | AB055895.1 | - | 2, 3 | 340 | ||
| BC089779.1 | - | 2, 3 | 329 | 7371893 | ||
| FQ210661.1 | - | 2, 3 | 340 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hrg| Gene Symbol: Hrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133428.2 | NP_596919.1 | 2, 3 | 340 | ||
| GenBank mRNA | AB055895.1 | - | 2, 3 | 340 | ||
| BC089779.1 | - | 2, 3 | 329 | 7371893 | ||
| FQ210661.1 | - | 2, 3 | 340 | |||