siRNA Details
| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004133.1 | NP_001004133.1 | 2, 3 | 726 | ||
| NM_133440.1 | NP_597684.1 | 2, 3 | 832 | |||
| NM_133441.1 | NP_597685.1 | 7, 8 | 1326 | |||
| NM_201350.1 | NP_958738.1 | 16, 17 | 2175 | |||
| XM_008760068.1 | XP_008758290.1 | 15, 16 | 2086 | |||
| GenBank mRNA | AF060116.1 | - | 2, 3 | 726 | ||
| AF141903.1 | - | 6, 7 | 818 | |||
| AF159048.1 | - | 6, 7 | 836 | |||
| AJ131899.1 | - | 2, 3 | 832 | |||
| AJ249562.1 | - | 7, 8 | 1326 | |||
| AY298755.1 | - | 16, 17 | 2175 | |||

Shank2
Genomic Map