siRNA Details
| Gene Symbol: Atp12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301664.1 | NP_001288593.1 | 1 | 518 | ||
| NM_133517.2 | NP_598201.2 | 2 | 377 | |||
| GenBank mRNA | M90398.1 | - | 2 | 301 | ||
| U94912.1 | - | 2 | 373 | |||
| U94913.1 | - | 1 | 517 | |||

Atp12a
Genomic Map