siRNA Details
| Gene Symbol: Tcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035237.1 | NP_001030314.1 | 8 | 652 | ||
| NM_133524.2 | NP_598208.2 | 8 | 652 | |||
| GenBank mRNA | AJ973227.1 | - | 8 | 652 | ||
| BC078750.1 | - | 8 | 580 | 7123064 | ||
| X54549.1 | - | 8 | 652 | |||
| X62323.1 | - | 8 | 556 | |||

Tcf3
Genomic Map