siRNA Details
| Gene Symbol: Tspan8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133526.1 | NP_598210.1 | 5 | 639 | ||
| GenBank mRNA | BC061785.1 | - | 5 | 495 | 5623788 | |
| FQ214867.1 | - | 5 | 547 | |||
| FQ215876.1 | - | 11 | ||||
| FQ216712.1 | - | 5 | 668 | |||
| FQ220073.1 | - | 11 | ||||
| FQ220093.1 | - | 11 | ||||
| FQ227464.1 | - | 5 | 656 | |||
| FQ229225.1 | - | 11 | ||||
| FQ229522.1 | - | 5 | 557 | |||
| Y13275.2 | - | 5 | 639 | |||

Tspan8
Genomic Map