siRNA Details
| Gene Symbol: Gpnmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133298.1 | NP_579832.1 | 11 | 1746 | ||
| GenBank mRNA | AF184983.2 | - | 11 | 1746 | ||
| BC061725.1 | - | 11 | 1689 | 5598392 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpnmb| Gene Symbol: Gpnmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133298.1 | NP_579832.1 | 11 | 1746 | ||
| GenBank mRNA | AF184983.2 | - | 11 | 1746 | ||
| BC061725.1 | - | 11 | 1689 | 5598392 | ||