siRNA Details
| Gene Symbol: Adarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133302.2 | NP_579836.1 | 1, 2 | 418 | ||
| GenBank mRNA | U74586.1 | - | 1, 2 | 419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adarb2| Gene Symbol: Adarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133302.2 | NP_579836.1 | 1, 2 | 418 | ||
| GenBank mRNA | U74586.1 | - | 1, 2 | 419 | ||