siRNA Details
| Gene Symbol: Capn8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133309.2 | NP_579843.2 | 6 | 838 | ||
| XM_006250363.2 | XP_006250425.2 | 6 | 1262 | |||
| GenBank mRNA | D14478.1 | - | 7 | 919 | ||
| D14479.1 | - | 6 | 838 | |||
| D14480.1 | - | 6 | 824 | |||

Capn8
Genomic Map