siRNA Details
Gene Symbol: Erg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133397.2 | NP_596888.1 | 5 | 700 | ||
XM_006248142.2 | XP_006248204.1 | 5 | 859 | |||
XM_006248143.2 | XP_006248205.1 | 5 | 859 | |||
XM_006248145.2 | XP_006248207.1 | 5 | 858 | |||
GenBank mRNA | AB031088.1 | - | 5 | 700 | ||
BC072519.1 | - | 5 | 680 | 7105647 | ||
FQ217228.1 | - | 5 | 719 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
