siRNA Details
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133420.1 | NP_596911.1 | 6 | 1503 | ||
XM_008770735.1 | XP_008768957.1 | 6 | 1345 | |||
GenBank mRNA | AY574253.1 | - | 5 | 1199 | ||
L10077.1 | - | 6 | 1503 |
