siRNA Details
| Gene Symbol: Cyb5r4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133427.3 | NP_596918.3 | 5 | 571 | ||
| XM_008766437.1 | XP_008764659.1 | 5 | 730 | |||
| GenBank mRNA | AF307840.1 | - | 4 | 330 | ||
| AY321370.1 | - | 4 | 330 | |||
| AY321371.1 | - | 4 | 330 | |||
| BC080240.1 | - | 5 | 549 | 7099950 | ||

Cyb5r4
Genomic Map