siRNA Details
| Gene Symbol: Gtf3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133541.1 | NP_598225.1 | 2 | 387 | ||
| XM_006230205.2 | XP_006230267.1 | 2 | 408 | |||
| GenBank mRNA | L28801.1 | - | 2 | 387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf3c1| Gene Symbol: Gtf3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133541.1 | NP_598225.1 | 2 | 387 | ||
| XM_006230205.2 | XP_006230267.1 | 2 | 408 | |||
| GenBank mRNA | L28801.1 | - | 2 | 387 | ||