siRNA Details
Gene Symbol: Cst6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133566.1 | NP_598250.1 | 2, 3 | 634 | ||
GenBank mRNA | AY046415.1 | - | 2, 3 | 634 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cst6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133566.1 | NP_598250.1 | 2, 3 | 634 | ||
GenBank mRNA | AY046415.1 | - | 2, 3 | 634 |