siRNA Details
| Gene Symbol: Adap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133567.2 | NP_598251.1 | 2 | 252 | ||
| GenBank mRNA | AF123047.1 | - | 2 | 179 | ||
| AJ007422.1 | - | 2 | 179 | |||
| AJ007616.1 | - | 2 | 179 | |||
| BC099775.1 | - | 2 | 252 | 7933317 | ||
| U51013.1 | - | 2 | 279 | |||

Adap1
Genomic Map