siRNA Details
Gene Symbol: Cdc25b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133572.1 | NP_598256.1 | 6, 7 | 811 | ||
GenBank mRNA | D16237.1 | - | 6, 7 | 811 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdc25b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133572.1 | NP_598256.1 | 6, 7 | 811 | ||
GenBank mRNA | D16237.1 | - | 6, 7 | 811 |