siRNA Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270864.1 | NP_001257793.1 | 2, 3 | 130 | ||
| NM_133583.2 | NP_598267.2 | 2, 3 | 148 | |||
| GenBank mRNA | AF334106.1 | - | 2, 3 | 112 | ||
| AJ426425.1 | - | 2, 3 | 161 | |||
| AJ426427.1 | - | 2, 3 | 124 | |||
| FM057759.1 | - | 2, 3 | 130 | |||

Ndrg2
Genomic Map