siRNA Details
| Gene Symbol: Kcne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133603.2 | NP_598287.1 | 2 | 284 | ||
| XM_006248023.2 | XP_006248085.1 | 2 | 710 | |||
| XM_008768554.1 | XP_008766776.1 | 2 | 520 | |||
| GenBank mRNA | AF071003.1 | - | 2 | 125 | ||
| BC089778.1 | - | 2 | 284 | 7301610 | ||

Kcne2
Genomic Map