siRNA Details
Gene Symbol: Taar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134328.1 | NP_599155.1 | 1 | 504 | ||
GenBank mRNA | AF421352.1 | - | 1 | 504 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
