siRNA Details
Gene Symbol: Ctsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134334.2 | NP_599161.2 | 2 | 267 | ||
GenBank mRNA | BC062032.1 | - | 2 | 267 | 5623627 | |
X54467.1 | - | 2 | 212 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ctsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134334.2 | NP_599161.2 | 2 | 267 | ||
GenBank mRNA | BC062032.1 | - | 2 | 267 | 5623627 | |
X54467.1 | - | 2 | 212 |