siRNA Details
| Gene Symbol: Cbwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133535.2 | NP_598219.2 | 3 | 293 | ||
| XM_006231191.2 | XP_006231253.1 | 3 | 381 | |||
| XM_006231192.2 | XP_006231254.1 | 3 | 381 | |||
| XM_006231193.2 | XP_006231255.1 | 3 | 381 | |||
| XM_008760303.1 | XP_008758525.1 | 3 | 381 | |||
| GenBank mRNA | AF353305.1 | - | 3 | 349 | ||
| BC086376.1 | - | 3 | 293 | 7135348 | ||

Cbwd1
Genomic Map